Assay Details
Gene Symbol: C17H1orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846038.4 | XP_851131.2 | 1 | 797 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C17H1orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846038.4 | XP_851131.2 | 1 | 797 | 94 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |