Assay Details
| Gene Symbol: PARVA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407845.1 | XP_022263553.1 | 9 - 10 | 806 | 111 | |
| XM_022407846.1 | XP_022263554.1 | 9 - 10 | 779 | 111 | ||
| XM_022407847.1 | XP_022263555.1 | 9 - 10 | 707 | 111 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PARVA
Genomic Map