Assay Details
| Gene Symbol: LOC479708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620930.3 | XP_005620987.1 | 773 | 154 | ||
| XM_022419526.1 | XP_022275234.1 | 774 | 154 | |||
| XM_022419527.1 | XP_022275235.1 | 774 | 154 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC479708
Genomic Map