Assay Details
| Gene Symbol: PBK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107472.2 | XP_013962947.1 | 5 - 6 | 542 | 98 | |
| XM_534564.6 | XP_534564.1 | 5 - 6 | 604 | 98 | ||
More Information
Set Membership: |
|

PBK
Genomic Map