Assay Details
| Gene Symbol: NCBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624963.2 | XP_005625020.1 | 11 | 1334 | 131 | |
| XM_022423490.1 | XP_022279198.1 | 10 | 1268 | 131 | ||
| XM_022423491.1 | XP_022279199.1 | 12 | 1352 | 131 | ||
| XM_849124.4 | XP_854217.2 | 11 | 1334 | 131 | ||
More Information
Set Membership: |
|

NCBP3
Genomic Map