Assay Details
| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622310.3 | XP_005622367.1 | 923 | 148 | ||
| XM_547387.5 | XP_547387.1 | 1656 | 148 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

YOD1
Genomic Map