Assay Details
| Gene Symbol: MNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423470.1 | XP_022279178.1 | 4 | 1215 | 125 | |
| XM_022423471.1 | XP_022279179.1 | 3 | 1189 | 125 | ||
More Information
Set Membership: |
|

MNT
Genomic Map