Assay Details
| Gene Symbol: ACVR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636801.3 | XP_005636858.1 | 3 | 585 | 119 | |
| XM_014108589.2 | XP_013964064.1 | 3 | 500 | 119 | ||
| XM_014108590.2 | XP_013964065.1 | 2 | 347 | 119 | ||
More Information
Set Membership: |
|

ACVR1B
Genomic Map