Assay Details
| Gene Symbol: PCDH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433053.4 | XP_003433101.1 | 2 | 1080 | 109 | |
| XM_005633954.3 | XP_005634011.1 | 2 | 1079 | 109 | ||
| XM_005633955.3 | XP_005634012.1 | 2 | 1079 | 109 | ||
| XM_022408012.1 | XP_022263720.1 | 2 | 1080 | 109 | ||
| XM_022408013.1 | XP_022263721.1 | 2 | 1080 | 109 | ||
| XM_022408014.1 | XP_022263722.1 | 2 | 1080 | 109 | ||
| XM_022408015.1 | XP_022263723.1 | 2 | 1080 | 109 | ||
| XM_843846.5 | XP_848939.1 | 2 | 1080 | 109 | ||
More Information
Set Membership: |
|

PCDH9
Genomic Map