Assay Details
| Gene Symbol: NMRAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420128.1 | XP_022275836.1 | 3 | 609 | 146 | |
| XM_547146.5 | XP_547146.3 | 3 | 604 | 146 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NMRAL1
Genomic Map