Assay Details
| Gene Symbol: ARHGAP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639524.3 | XP_005639581.1 | 11 - 12 | 2061 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARHGAP31| Gene Symbol: ARHGAP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639524.3 | XP_005639581.1 | 11 - 12 | 2061 | 72 | |
Set Membership: |
|