Assay Details
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639506.2 | XP_005639563.1 | 13 - 14 | 2324 | 146 | |
XM_005639507.2 | XP_005639564.1 | 12 - 13 | 2285 | 146 | ||
XM_014110273.2 | XP_013965748.1 | 12 - 13 | 2175 | 146 | ||
XM_022414004.1 | XP_022269712.1 | 12 - 13 | 2057 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
