Assay Details
| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413452.1 | XP_022269160.1 | 8 - 9 | 731 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

METTL14| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413452.1 | XP_022269160.1 | 8 - 9 | 731 | 111 | |
Set Membership: |
|