Assay Details
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022409481.1 | XP_022265189.1 | 6 - 7 | 516 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022409481.1 | XP_022265189.1 | 6 - 7 | 516 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |