Assay Details
| Gene Symbol: TRIB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634904.3 | XP_005634961.1 | 2 - 3 | 474 | 114 | |
| XM_542943.5 | XP_542943.2 | 2 - 3 | 348 | 114 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TRIB3
Genomic Map