Assay Details
| Gene Symbol: MYOZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048095.1 | NP_001041560.1 | 2 - 3 | 248 | 140 | |
| XM_005618876.2 | XP_005618933.1 | 3 - 4 | 368 | 140 | ||
| GenBank mRNA | DQ143043.1 | - | 2 - 3 | 248 | 140 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MYOZ1
Genomic Map