Assay Details
| Gene Symbol: GPBP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629053.3 | XP_005629110.1 | 3 - 4 | 1313 | 81 | |
| XM_005629054.3 | XP_005629111.1 | 3 - 4 | 1273 | 81 | ||
| XM_022427998.1 | XP_022283706.1 | 3 - 4 | 1160 | 81 | ||
| XM_532596.6 | XP_532596.2 | 3 - 4 | 1241 | 81 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GPBP1L1
Genomic Map