Assay Details
Gene Symbol: ZFAND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118409.2 | XP_013973884.1 | 4 | 324 | 130 | |
XM_022426011.1 | XP_022281719.1 | 3 | 528 | 130 | ||
XM_538897.6 | XP_538897.3 | 3 | 294 | 130 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
