Assay Details
| Gene Symbol: LAMTOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639229.3 | XP_005639286.1 | 5 - 6 | 544 | 115 | |
| XM_022413535.1 | XP_022269243.1 | 5 - 6 | 463 | 115 | ||
| XM_857268.5 | XP_862361.1 | 5 - 6 | 435 | 115 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LAMTOR3
Genomic Map