Assay Details
| Gene Symbol: FAM204A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109083.2 | XP_013964558.1 | 6 - 7 | 636 | 64 | |
| XM_535027.6 | XP_535027.4 | 6 - 7 | 691 | 64 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FAM204A
Genomic Map