Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408561.1 | XP_022264269.1 | 3 - 4 | 715 | 163 | |
| XM_542751.6 | XP_542751.3 | 3 - 4 | 715 | 163 | ||
More Information
Set Membership: |
|

AZI2
Genomic Map