Assay Details
| Gene Symbol: RFX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640056.4 | XP_003640104.1 | 9 - 10 | 1443 | 92 | |
| XM_022412529.1 | XP_022268237.1 | 9 - 10 | 1446 | 92 | ||
| XM_022412530.1 | XP_022268238.1 | 9 - 10 | 1226 | 92 | ||
More Information
Set Membership: |
|

RFX7
Genomic Map