Assay Details
| Gene Symbol: ADIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637369.3 | XP_005637426.1 | 3 - 4 | 601 | 98 | |
| XM_005637370.3 | XP_005637427.1 | 3 - 4 | 536 | 98 | ||
| XM_022411304.1 | XP_022267012.1 | 3 - 4 | 394 | 98 | ||
| XM_863038.5 | XP_868131.1 | 3 - 4 | 595 | 98 | ||
More Information
Set Membership: |
|

ADIPOR2
Genomic Map