Assay Details
| Gene Symbol: NAA40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631557.3 | XP_005631614.2 | 2 - 3 | 214 | 120 | |
| XM_014121321.2 | XP_013976796.1 | 2 - 3 | 126 | 120 | ||
| XM_022405756.1 | XP_022261464.1 | 2 - 3 | 223 | 120 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NAA40
Genomic Map