Assay Details
| Gene Symbol: SYNPO2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638990.4 | XP_003639038.1 | 4 | 1831 | 135 | |
| XM_005619036.3 | XP_005619093.1 | 2 | 1737 | 135 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SYNPO2L
Genomic Map