Assay Details
| Gene Symbol: PTGES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131050.1 | NP_001124522.1 | 1 | 324 | 277 | |
| XM_005625033.3 | XP_005625090.1 | 1 | 270 | 172 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTGES2
Genomic Map