Assay Details
| Gene Symbol: TM2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434355.2 | XP_003434403.1 | 5 | 614 | 118 | |
| XM_005618279.1 | XP_005618336.1 | 6 | 707 | 118 | ||
| XM_022416832.1 | XP_022272540.1 | 7 | 833 | 118 | ||
| XM_022416833.1 | XP_022272541.1 | 6 | 740 | 118 | ||
| XM_022416834.1 | XP_022272542.1 | 7 | 849 | 118 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TM2D3
Genomic Map