Assay Details
| Gene Symbol: COASY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624450.3 | XP_005624507.1 | 1324 | 100 | ||
| XM_005624451.2 | XP_005624508.1 | 986 | 100 | |||
| XM_537637.6 | XP_537637.2 | 1052 | 100 | |||
More Information
Set Membership: |
|

COASY
Genomic Map