Assay Details
| Gene Symbol: SEC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632167.2 | XP_005632224.1 | 1351 | 100 | ||
| XM_845344.4 | XP_850437.1 | 1 | 972 | 100 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEC13
Genomic Map