Assay Details
| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313853.1 | NP_001300782.1 | 8 | 1241 | 126 | |
| GenBank mRNA | AB755429.1 | - | 8 | 1241 | 126 | |
| JX629803.1 | - | 8 | 1241 | 126 | ||
| KC165685.1 | - | 8 | 1241 | 126 | ||
| KJ623742.1 | - | 8 | 1241 | 126 | ||
More Information
Set Membership: |
|

NECTIN4
Genomic Map