Assay Details
| Gene Symbol: RAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405629.1 | XP_022261337.1 | 3 - 4 | 369 | 112 | |
| XM_847452.5 | XP_852545.1 | 3 - 4 | 307 | 112 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB1B
Genomic Map