Assay Details
| Gene Symbol: RAMAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618394.3 | XP_005618451.1 | 4 | 336 | 123 | |
| XM_005618395.3 | XP_005618452.1 | 3 | 414 | 123 | ||
| XM_005618396.3 | XP_005618453.1 | 3 | 366 | 123 | ||
| XM_536205.6 | XP_536205.1 | 4 | 367 | 123 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAMAC
Genomic Map