Assay Details
| Gene Symbol: COQ5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636306.3 | XP_005636363.1 | 4 | 584 | 125 | |
| XM_022410440.1 | XP_022266148.1 | 3 | 535 | 125 | ||
| XM_849175.5 | XP_854268.1 | 4 | 685 | 125 | ||
More Information
Set Membership: |
|

COQ5
Genomic Map