Assay Details
| Gene Symbol: LZTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637632.3 | XP_005637689.1 | 2 - 3 | 585 | 185 | |
| XM_014108974.2 | XP_013964449.1 | 1 - 2 | 2723 | 185 | ||
| XM_014108975.2 | XP_013964450.1 | 3 - 4 | 1111 | 185 | ||
| XM_022412116.1 | XP_022267824.1 | 4 - 5 | 988 | 185 | ||
More Information
Set Membership: |
|

LZTS2
Genomic Map