Assay Details
| Gene Symbol: USP38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405875.1 | XP_022261583.1 | 990 | 98 | ||
| XM_022405876.1 | XP_022261584.1 | 990 | 98 | |||
| XM_022405877.1 | XP_022261585.1 | 990 | 98 | |||
| XM_533279.6 | XP_533279.2 | 990 | 98 | |||
More Information
Set Membership: |
|

USP38
Genomic Map