Assay Details
| Gene Symbol: LOC490877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623872.3 | XP_005623929.1 | 3 - 4 | 737 | 102 | |
| XM_547999.6 | XP_547999.4 | 3 - 4 | 736 | 102 | ||
More Information
Set Membership: |
|

LOC490877
Genomic Map