Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631444.3 | XP_005631501.1 | 3 | 796 | 143 | |
| XM_540838.5 | XP_540838.2 | 3 | 2770 | 143 | ||
More Information
Set Membership: |
|

GAL3ST3
Genomic Map