Assay Details
| Gene Symbol: KNSTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640036.4 | XP_003640084.1 | 7 - 8 | 906 | 165 | |
| XM_014109597.2 | XP_013965072.1 | 6 - 7 | 859 | 165 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KNSTRN
Genomic Map