Assay Details
| Gene Symbol: FHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119711.2 | XP_013975186.1 | 7 - 8 | 1129 | 109 | |
| XM_539768.5 | XP_539768.3 | 7 - 8 | 1140 | 109 | ||
More Information
Set Membership: |
|

FHDC1
Genomic Map