Assay Details
| Gene Symbol: WFIKKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_547215.6 | XP_547215.4 | 2 | 1766 | 171 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFIKKN1| Gene Symbol: WFIKKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_547215.6 | XP_547215.4 | 2 | 1766 | 171 | |
Set Membership: |
|