Assay Details
| Gene Symbol: DYNLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624719.2 | XP_005624776.1 | 2 - 3 | 296 | 138 | |
| XM_014116688.2 | XP_013972163.1 | 2 - 3 | 244 | 138 | ||
| XM_022423187.1 | XP_022278895.1 | 2 - 3 | 235 | 138 | ||
| XM_022423189.1 | XP_022278897.1 | 2 - 3 | 282 | 138 | ||
| XM_022423190.1 | XP_022278898.1 | 2 - 3 | 257 | 138 | ||
| XM_861763.4 | XP_866856.1 | 2 - 3 | 203 | 138 | ||
| XM_861812.5 | XP_866905.1 | 2 - 3 | 286 | 138 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DYNLL2
Genomic Map