Assay Details
| Gene Symbol: RBBP8NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409231.1 | XP_022264939.1 | 8 - 9 | 1203 | 95 | |
| XM_022409232.1 | XP_022264940.1 | 8 - 9 | 1198 | 95 | ||
More Information
Set Membership: |
|

RBBP8NL
Genomic Map