Assay Details
| Gene Symbol: SPSB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634513.3 | XP_005634570.1 | 4 | 1045 | 228 | |
| XM_022408697.1 | XP_022264405.1 | 3 | 951 | 147 | ||
More Information
Set Membership: |
|

SPSB4
Genomic Map