Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406560.1 | XP_022262268.1 | 8 | 1522 | 149 | |
| XM_022406561.1 | XP_022262269.1 | 8 | 1522 | 149 | ||
| XM_022406562.1 | XP_022262270.1 | 8 | 1535 | 149 | ||
| XM_022406563.1 | XP_022262271.1 | 7 | 1369 | 149 | ||
| XM_022406564.1 | XP_022262272.1 | 7 | 1276 | 149 | ||
More Information
Set Membership: |
|

ASB14
Genomic Map