Assay Details
| Gene Symbol: RAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619405.3 | XP_005619462.1 | 2 - 3 | 577 | 140 | |
| XM_022417935.1 | XP_022273643.1 | 5 - 6 | 977 | 140 | ||
| XM_022417936.1 | XP_022273644.1 | 6 - 7 | 1048 | 140 | ||
| XM_022417937.1 | XP_022273645.1 | 3 - 4 | 647 | 140 | ||
| XM_536505.6 | XP_536505.3 | 2 - 3 | 425 | 140 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAD1
Genomic Map