Assay Details
| Gene Symbol: MED22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625143.3 | XP_005625200.1 | 3 - 4 | 372 | 135 | |
| XM_005625144.3 | XP_005625201.1 | 3 - 4 | 354 | 135 | ||
| XM_022423633.1 | XP_022279341.1 | 3 - 4 | 247 | 135 | ||
| XM_537799.6 | XP_537799.3 | 3 - 4 | 420 | 135 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MED22
Genomic Map