Assay Details
| Gene Symbol: BORCS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_535001.6 | XP_535001.3 | 2 - 3 | 304 | 206 | |
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length greater than or equal to 101  Recommended Assay  Canis familiaris  Probe spans exons | 
 Back To Top
 Back To Top
 BORCS7
BORCS7 Genomic Map
Genomic Map Back To Top
 Back To Top