Assay Details
| Gene Symbol: SYT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622720.3 | XP_005622777.1 | 4 | 1746 | 137 | |
| XM_846821.5 | XP_851914.2 | 4 | 1588 | 137 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SYT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622720.3 | XP_005622777.1 | 4 | 1746 | 137 | |
| XM_846821.5 | XP_851914.2 | 4 | 1588 | 137 | ||
Set Membership: |
|