Assay Details
| Gene Symbol: PXYLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634516.3 | XP_005634573.1 | 7 | 1409 | 118 | |
| XM_005634517.3 | XP_005634574.1 | 7 | 1361 | 118 | ||
| XM_005634518.3 | XP_005634575.1 | 7 | 1715 | 118 | ||
| XM_005634519.3 | XP_005634576.1 | 5 | 1191 | 118 | ||
| XM_022408699.1 | XP_022264407.1 | 7 | 1409 | 118 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PXYLP1
Genomic Map