Assay Details
| Gene Symbol: CFAP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119618.2 | XP_013975093.1 | 8 - 9 | 1649 | 176 | |
| XM_014119619.2 | XP_013975094.1 | 8 - 9 | 1652 | 176 | ||
| XM_014119620.2 | XP_013975095.1 | 7 - 8 | 1496 | 176 | ||
| XM_022428072.1 | XP_022283780.1 | 6 - 7 | 1091 | 176 | ||
More Information
Set Membership: |
|

CFAP57
Genomic Map